

Hydrofluoric acid 49.5-50.5%, Semiconductor grade (Honeywell 17736)
Catalog # HONC40213H-5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Assay:49.5 - 50.5 %
- Residue on ignition (SO₄):Max. 5 ppm
- Silver (Ag):Max. 0.02 ppm
- Aluminium (Al):Max. 0.05 ppm
- Arsenic (As):Max. 0.05 ppm
- Gold (Au):Max. 0.02 ppm
- Boron (B):Max. 0.05 ppm
- Barium (Ba):Max. 0.1 ppm
- Beryllium (Be):Max. 0.01 ppm
- Bismuth (Bi):Max. 0.02 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.01 ppm
- Cobalt (Co):Max. 0.01 ppm
- Chromium (Cr):Max. 0.01 ppm
- Copper (Cu):Max. 0.01 ppm
- Iron (Fe):Max. 0.1 ppm
- Gallium (Ga):Max. 0.02 ppm
- Indium (In):Max. 0.02 ppm
- Potassium (K):Max. 0.1 ppm
- Lithium (Li):Max. 0.02 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.01 ppm
- Molybdenum (Mo):Max. 0.01 ppm
- Sodium (Na):Max. 0.2 ppm
- Nickel (Ni):Max. 0.01 ppm
- Lead (Pb):Max. 0.02 ppm
- Platinum (Pt):Max. 0.02 ppm
- Antimony (Sb):Max. 0.01 ppm
- Tin (Sn):Max. 0.02 ppm
- Strontium (Sr):Max. 0.02 ppm
- Titanium (Ti):Max. 0.1 ppm
- Thallium (Tl):Max. 0.02 ppm
- Vanadium (V):Max. 0.01 ppm
- Zinc (Zn):Max. 0.05 ppm
- Zirconium (Zr):Max. 0.01 ppm
- Chloride (Cl):Max. 5 ppm
- Hexafluorosilicic acid (H₂SiF₆):Max. 50 ppm
- Nitrate (NO₃):Max. 3 ppm
- Phosphate (PO₄):Max. 0.5 ppm
- Sulfate (SO₄):Max. 1 ppm
- Sulfite (SO₃):Max. 2 ppm
Specification Test Results
About this item
:
:20.01 g/mol
Boiling Pt:105 °C (1.013 hPa)
Melting Pt:–35 °C
Density:1.170 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-634-8
UN:1790
ADR:8,II
:GHS05
:GHS06
CAS Number:7664-39-3
Formula:HF
MDL Number:MFCD00011324