Methanol with 0,1% (v/v) acetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
HONC34672-4X2.5L
:
- Pack type:Glass bottle
- Pk:10 L
- Seq:5114
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
- Pk Info:4x 2.5 L
- Assay (GC):min. 99.5 %
- Calcium (Ca):max. 0.5 ppm
- Potassium (K):max. 0.5 ppm
- Magnesium (Mg):max. 0.5 ppm
- Sodium (Na):max. 2 ppm
- Transmittance at 210 nm:min. 5 %
- Transmittance at 230 nm:min. 50 %
- Transmittance from 254 nm:min. 95 %
- HPLC Gradient at 254 nm:max. 10 mAU
- LC-MS application:Complying
:
:68 °C
:0.79 g/cm³ (20 °C)
:20 °C
:Ambient
:200-659-6
:1230
:3,II
:GHS02
:GHS08
:67-56-1
:64-19-7
:MFCD00004595