Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Ethyl acetate ≥99.0% (by GC), Laboratory Reagent
    
 
undefined
 
 
Ethyl acetate ≥99.0% (by GC), Laboratory Reagent
    

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Assay (GC):
    Min. 99.0 %
  • Free acid (as CH₃COOH):
    Max. 0.003 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Non-volatile matter:
    Max. 0.003 %
  • Explosion Limit:
    2.2-11.5 %, 38 °F
  • Refractive Index:
    n20/D 1.3720 (lit.)
  • Autoignition Temperature:
    801 °F

 

 

Frequently Bought Together