
Ethanol absolute, anhydrous ≥99.8% (by GC), AR®, Macron Fine Chemicals™
MACR6777-16
:
- Pk:2,5 L
- Seq:5096
- Assay (GC):≥99.8%
- Water (KF):≤0.2%
- Titrable Acid (meq/g):≤0.0002
- Residue after Evaporation:≤0.001%
- Aldehydes and ketones (as C₃H₆O) :≤0.001%
- Higher alcohols (as amyl alcohol):≤0.015%
- Methyl Alcohol:≤0.01%
- Time for reducing KMnO4 solution:≥10 mins
- Substances Darkened by H₂SO₄:Passes test
- Carbonyl compounds (as CO):≤0.003%
:
:46.07 g/mol
:Ambient
:1170
:3,II
:GHS02
:64-17-5
:C₂H₅OH
:MFCD00003568