Order Entry
Belgium
ContactUsLinkComponent
VWR® IsoFreeze®, PCR and qPCR Racks, Colour Change
VWR® IsoFreeze®, PCR and qPCR Racks, Colour Change
Catalog # 732-3498
Supplier:  VWR Collection
VWR® IsoFreeze®, PCR and qPCR Racks, Colour Change
Catalog # 732-3498
Supplier:  VWR Collection

Specifications

  • Colour:
    Lilac
  • Description:
    PCR racks, colour change: purple to pink
  • Seq:
    02
  • Pk:
    2

Specifications

About this item

Iso-freeze PCR racks are specially designed for Qiagen® and Corbett Rotor-Gene® tubes.

  • Maintain a sample temperature of less than 4 °C for almost 4 hours (with lid on rack) when used at room temperature
  • Gradual fade in colour changes indicates slowly rising sample temperatures
  • It undergoes a dramatic change to a contrasting colour at 7 °C
  • Blue rack colour indicates safe sample temperature

Tube and plate capacity:
PCR: 96×0,1 and 0,2 ml tubes and strip tube, plates up to 96-well.
qPCR: 64×0,1 ml Corbett rotor-gene tubes, 16×0,2 ml tubes or strips, and 4×0,5 ml tubes.

Dimensions: 141×99×44 mm