Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®, Support Stand, Rectangular Base, Cast Iron

VWR®, Support Stand, Rectangular Base, Cast Iron

Rectangular base support stand, cast iron, 254×203 mm, Black
Catalog # 444-0918
Supplier:   VWR Collection
VWR®, Support Stand, Rectangular Base, Cast Iron
Catalog # 444-0918
Supplier:  VWR Collection
VWR®, Support Stand, Rectangular Base, Cast Iron

Rectangular base support stand, cast iron, 254×203 mm, Black

Price
184,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Cast iron support stand bases are constructed with black enamel finish for chemical resistance and durability.

It features a built-in support rod holder with locking knob that accepts a 13 mm diameter support rod.

Ordering information: Support rods are not included.

Specifications
  • Colour:
    Black
  • Description:
    Rectangular base support stand, cast iron
  • L×W:
    254×203 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.