ContactUsLinkComponent
Trace Metals 1-WP, certified reference material, Supelco®
Trace Metals 1-WP, certified reference material, Supelco®
  QC1132-20ML
 :  Merck
 :  
Trace Metals 1-WP, certified reference material, Supelco®
  QC1132-20ML
 :  Merck
 :  
  • Matrix:
    5% HNO₃
  • Pack type:
    Ampoule
  • Pk:
    20 mL
  • Description:
    Trace Metals 1-WP
Analytes:
Aluminium; Arsenic; Beryllium;
Cadmium; Chromium; Cobalt; Copper;
Iron; Lead; Lithium;
Manganese; Mercury; Nickel;
Selenium; Vanadium; Zinc
 : This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.