Avantor® Hichrom, ValvTool
Catalog # HI-199
Supplier: Avantor
Specifications
- Description:ValvTool
- Environmentally Preferable:
- Use with head style:1/4"
- Pk:1
Specifications
About this item
This Valvtool is a uniquely designed slotted wrench which provides easy access to many hard-to-reach areas.
It enables fittings, sample loops and most other ¼" stainless steel and PEEK HPLC fittings to be more easily tightened or loosened.