Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tubes
Test Tubes
VWR®, Sample tubes, soda glass
VWR®, Sample tubes, soda glass
VWR®, Sample tubes, soda glass
406047200.
:
VWR Chemicals
VWR®, Sample tubes, soda glass
406047200.
:
VWR Chemicals
Ext.Ø:
1,0 mm
Description:
Sample tube with bore
Length:
90 mm
Material:
Soda glass
Seq:
01
Pk:
1.000
Tubes are used for lymph blood samples.