VWR®, Centrifuge Tubes, Conical Bottom, with Flat Cap
Supplier: VWR Collection
Certificates
About this item
Individually wrapped sterile centrifuge tube.
- Flat cap with sealing ring
- During the freezing process, it is better to use plastic rack
- During high-speed centrifugation, it is better to use suitable rotor adapters
Specifications
- Catalog No:
- 525-1437
- 525-1311
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M