Orders LinkContactUsLinkComponent
Brushes for test tubes
Brushes for test tubes
Brushes for test tubes
Certificates
Brushes for test tubes
2 Options Available Below
Product Details & Documents
Brushes with natural bristles and zinc plated wire handle.
Product Family Options
Product Information
  • Bristle material
    Brush head length
    Brush head Ø
    Handle material
    Total length
    Availability
    Price
  • REIT12200301
    Test tube brush, with fallow bristles
    Test tube brush, with fallow bristles
    Hog bristle
    60 mm
    10 mm
    Wire
    190 mm
    Discontinued
  • REIT12210301
    Test tube brush, with fallow bristles
    Test tube brush, with fallow bristles
    Hog bristle
    70 mm
    16 mm
    Wire
    190 mm
    Discontinued
  • REIT12220301
    Test tube brush, with fallow bristles
    Test tube brush, with fallow bristles
    Hog bristle
    80 mm
    18 mm
    Wire
    220 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 70,50 €
    70,50 €
    1 * 10 ST
       70,50 €
  • REIT12230301
    Test tube brush, with fallow bristles
    Test tube brush, with fallow bristles
    Hog bristle
    95 mm
    21 mm
    Wire
    230 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,30 €
    11,30 €
    1 * 1 ST
       11,30 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.