Order Entry
Belgium
Orders LinkContactUsLinkComponent
Baret cap

Baret cap

White
Catalog # HOIT114510_U
Baret cap
Catalog # HOIT114510_U
Baret cap
Baret cap

White

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

This baret cap is made from polypropylene.

  • With Peak

Weight: Peak - 90 g/m², Cap - 35 g/m²

Colour: White.

Packaging: 100 pieces/bag; 10 bags/case.

Specifications
  • Colour:
    White
  • Cat. No.:
    HOIT114510_U
  • Weight:
    35 g/m²