Order Entry
Belgium
ContactUsLinkComponent
 
Reversed phase test mix, in methanol: water (3:2) (varied conc.), certified reference material, Supelco®
Catalog # 58278.
Supplier:  Merck
CAS Number:  
Reversed phase test mix, in methanol: water (3:2) (varied conc.), certified reference material, Supelco®
Catalog # 58278.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    Reversed Phase Test Mix, in methanol: water (3:2) (varied conc.)
  • Storage temperature:
    Ambient
  • Pk Info:
    ampule of 1 mL

Specifications

About this item

Analytes:

Acetophenone 7 μg/ml; Benzene 750 μg/ml; Toluene 775 μg/ml; Uracil 7 μg/ml

Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.