Order Entry
Belgium
ContactUsLinkComponent
Reversed phase test mix 2, certified reference material, Supelco®
Reversed phase test mix 2, certified reference material, Supelco®
Catalog # 47641-U
Supplier:  Merck
CAS Number:  
Reversed phase test mix 2, certified reference material, Supelco®
Catalog # 47641-U
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    in acetonitrile: water (varied conc.) solution
  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    Reversed Phase Test Mix 2
  • Storage temperature:
    2 - 8 °C
  • Pk Info:
    ampule of 1 mL

Specifications

About this item

Analytes:

N,N-Diethyl-m-toluamide 600 μg/ml; Phenol 700 μg/ml; Toluene 4000 μg/ml; Uracil 5 μg/ml

Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.