Order Entry
Belgium
ContactUsLinkComponent
Poly(lauryl methacrylate) solution, Mw 570000 (Typical), average Mn 290000 (Typical), analytical standard, Supelco®
Poly(lauryl methacrylate) solution, Mw 570000 (Typical), average Mn 290000 (Typical), analytical standard, Supelco®
Catalog # 182192-5G
Supplier:  Merck
CAS Number:  
undefined
Poly(lauryl methacrylate) solution, Mw 570000 (Typical), average Mn 290000 (Typical), analytical standard, Supelco®
Catalog # 182192-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    25 wt. % in toluene
  • Pk:
    5 g
  • Standards:
    Analytical standard
  • CAS No.:
    25719-52-2
  • Description:
    Poly(lauryl methacrylate) solution, Mw 570000 (Typical), average Mn 290000 (Typical)

Specifications

About this item