Order Entry
Belgium
ContactUsLinkComponent
Poly(butyl acrylate) solution, Mw 60000 (Typical), average Mn 20000 (Typical), analytical standard, Supelco®
Poly(butyl acrylate) solution, Mw 60000 (Typical), average Mn 20000 (Typical), analytical standard, Supelco®
Catalog # 181412-10G
Supplier:  Merck
CAS Number:  
undefined
Poly(butyl acrylate) solution, Mw 60000 (Typical), average Mn 20000 (Typical), analytical standard, Supelco®
Catalog # 181412-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    25 wt. % in toluene
  • Pk:
    10 g
  • Standards:
    Analytical standard
  • CAS No.:
    9003-49-0
  • Description:
    Poly(butyl acrylate) solution, Mw 60000 (Typical), average Mn 20000 (Typical)

Specifications

About this item