Order Entry
Belgium
ContactUsLinkComponent
Intermediate polar column test mix, certified reference material, Supelco®
Intermediate polar column test mix, certified reference material, Supelco®
Catalog # 47301.
Supplier:  Merck
CAS Number:  
Intermediate polar column test mix, certified reference material, Supelco®
Catalog # 47301.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    500 mug/mL in dichloromethane
  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    Intermediate Polar Column Test Mix
  • Pk Info:
    ampule of 2 mL

Specifications

About this item

Analytes:

Decane; 2,6-Dimethylaniline; 2,6-Dimethylphenol; Dodecane; 1-Octanol; 2-Octanone; Tetradecane; Tridecane; Undecane

Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.