Order Entry
Belgium
ContactUsLinkComponent
HPLC gradient system diagnostics mix, in methanol: water (3:2) (varied concentration), certified reference material, Supelco®
HPLC gradient system diagnostics mix, in methanol: water (3:2) (varied concentration), certified reference material, Supelco®
Catalog # 48271.
Supplier:  Merck
CAS Number:  
HPLC gradient system diagnostics mix, in methanol: water (3:2) (varied concentration), certified reference material, Supelco®
Catalog # 48271.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    6 mL
  • Standards:
    Certified reference material
  • Description:
    HPLC Gradient System Diagnostics Mix, in methanol: water (3:2) (varied conc.)
  • Pk Info:
    pkg of 6 × 1 ml

Specifications

About this item

This formulation is designed for evaluating how reliably a chromatographic system is providing such fundamentally important parameters as flow rate, proportioning, and mixing.

Analytes:

Butyl 4-hydroxybenzoate; Ethyl 4-hydroxybenzoate; Heptyl paraben; Methyl 4-hydroxybenzoate; Phenol; Propyl 4-hydroxybenzoate; Uracil

Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.