Order Entry
Belgium
ContactUsLinkComponent
 
Phosphate standard solution, concentrate, 1,000 mg PO₄, Titrisol®, Supelco®
  1.09870.0001
 :  Merck
 :  
undefined
Phosphate standard solution, concentrate, 1,000 mg PO₄, Titrisol®, Supelco®
  1.09870.0001
 :  Merck
 :  

 

  • Concentration:
    1000 mg/l
  • Pk:
    1 Ampoul
  • CAS No.:
    7664-38-2
  • Description:
    Phosphate standard, Titrisol®
  • M-Clarity quality level:
    MQ100
  • Contents:
    H3PO4 in water