Order Entry
Belgium
ContactUsLinkComponent
Melting point standards, sodium acetate anhydrous, traceable to primary standards, Supelco®
Melting point standards, sodium acetate anhydrous, traceable to primary standards, Supelco®
Catalog # 42123-5G
Supplier:  Merck
CAS Number:  
undefined
Melting point standards, sodium acetate anhydrous, traceable to primary standards, Supelco®
Catalog # 42123-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 g
  • Description:
    Sodium acetate
  • Melting pt.:
    329,4 °C
  • Accuracy:
    ± 0,4 °C

Specifications

About this item

Mettler-Toledo Calibration substance ME 30130599, Sodium acetate anhydrous, tracable to primary standards (LGC).

Suitable for dropping point calibration.