Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, Iopromide
Supelco® Organic Reference Standard, Iopromide
Catalog # 38701-25MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Iopromide
Catalog # 38701-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    73334-07-3
  • Description:
    Iopromide
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00867924
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    791,11
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS09
Caution: P273 - P501