Supelco® PESTANAL® Organic Reference Standard, Pyriproxyfen
Catalog # 34174-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:95737-68-1
- Description:Pyriproxyfen
- Storage conditions:2...30 °C
- Hazard class description:H410
- Shipping conditions:STD
- MDL No.:MFCD01691614
- M-Clarity quality level:MQ100
- Synonym(s):2-[1-Methyl-2-(4-phenoxyphenoxy)ethoxy]pyridine
- Molecular weight:321,37
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS09
Caution: P273 - P391 - P501