Order Entry
Belgium
ContactUsLinkComponent
Microcystin-RR solution, analytical standard, Supelco®
Microcystin-RR solution, analytical standard, Supelco®
Catalog # 33577-1ML
Supplier:  Merck
CAS Number:  
undefined
Microcystin-RR solution, analytical standard, Supelco®
Catalog # 33577-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 mug/mL in methanol
  • Pk:
    1 mL
  • Standards:
    analytical standard
  • CAS No.:
    111755-37-4
  • Description:
    Microcystin-RR solution
  • M-Clarity quality level:
    MQ100
  • Description:
    Microcystin-RR solution

Specifications

About this item