0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Standards
Organic Standards
Triclabendazole, analytical standard, VETRANAL™, Supelco®
Triclabendazole, analytical standard, VETRANAL™, Supelco®
32802-100MG
:
Merck
:
Triclabendazole, analytical standard, VETRANAL™, Supelco®
32802-100MG
:
Merck
:
Pk:
100 mg
Standards:
analytical standard
CAS No.:
68786-66-3
Description:
Triclabendazole
M-Clarity quality level:
MQ100
Description:
Triclabendazole