Supelco® VETRANAL® Organic Reference Standard, Terconazole
32355-100MG
: Merck
:
- Pk:100 mg
- Standards:VETRANAL®, Analytical standard
- CAS No.:67915-31-5
- Description:Terconazole
- Storage conditions:2...30 °C
- Hazard class description:H315 - H319 - H335 - H413
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):(+-)-1-{4-[cis-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-ylmethoxy]phenyl}-4-isopropylpiperazine; Triaconazole
- Molecular weight:532,46
- Flash point:Not applicable
- GHS signal: GHS07
: P261 - P305 + P351 + P338