Order Entry
Belgium
Orders LinkContactUsLinkComponent
Phenol red solution, special quality indicator

Phenol red solution, special quality indicator

50 mL
Catalog # HACH2657512
Supplier:   Hach
Phenol red solution, special quality indicator
Catalog # HACH2657512
Supplier:  Hach
CAS Number:  143-74-8
Phenol red solution, special quality indicator
Phenol red solution, special quality indicator

50 mL

CAS Number:  143-74-8
Price
26,90 €
Log in for availability
1 * 50 mL
Product Details & Documents
Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Formula:C₁₉H₁₄O₅S
  • MDL Number:MFCD00003552
  • Molecular Weight:354.38 g/mol
Specifications

Specifications

  • Pk:
    50 mL

Specifications