ContactUsLinkComponent
Cervical brushes, Cytotak™ Transwab®, labelled
Cervical brushes, Cytotak™ Transwab®, labelled
Catalog # 720-0136
undefined
Cervical brushes, Cytotak™ Transwab®, labelled
Catalog # 720-0136
Specifications
  • Handle material:
    Plastic
  • Description:
    Sterile, in labelled tube
  • Seq:
    01
  • Total length:
    150 mm
  • Environmentally Preferable:
  • Pk:
    100
Specifications
About this item
Nylon-bristle cytology sampling brushes for cervical smears.
  • Sterile, packed in labelled tube
  • Plastic shaft is non toxic and safe to use
  • Tapered spiral configuration assists access to narrow opening
  • Gentle rotation creates a brushing action against skin surface for enhanced recovery of cellular material
Shaft/head: Plastic/nylon
Packaging: Individually packed.