Nitrite nitrogen standard for IC, certified reference material, TraceCERT®, Supelco®
N as nitrite in water (pH ~11), 1000 mg/l, 100 mL
Supplier: Merck
Nitrite nitrogen standard for IC, certified reference material, TraceCERT®, Supelco®
Catalog # 36427-100ML
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Concentration:1000 mg/l
- Pk:100 mL
- Description:N as nitrite in water (pH ~11)
Specifications
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



