Order Entry
Belgium
ContactUsLinkComponent
Nitrite ion standard solution, for ion-selective electrodes, Supelco®
Nitrite ion standard solution, for ion-selective electrodes, Supelco®
Catalog # 72586-100ML
Supplier:  Merck
CAS Number:  
undefined
Nitrite ion standard solution, for ion-selective electrodes, Supelco®
Catalog # 72586-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    0,1 M NO2-
  • Pk:
    100 mL
  • Description:
    Sodium nitrite in water
  • M-Clarity quality level:
    MQ100

Specifications

About this item

Prepared with NaNO₂ and H₂O.

Caution: Storage Temperature: 2 to 8 °C