Order Entry
Belgium
Orders LinkContactUsLinkComponent
Lactate standard for IC, TraceCERT®, Supelco®

Lactate standard for IC, TraceCERT®, Supelco®

Lactate in water, 1000 mg/l, 100 mL
Catalog # 07096-100ML
Supplier:   Merck
Lactate standard for IC, TraceCERT®, Supelco®
Catalog # 07096-100ML
Supplier:  Merck
CAS Number:  
Lactate standard for IC, TraceCERT®, Supelco®

Lactate in water, 1000 mg/l, 100 mL

CAS Number:  
Price
61,10 €
Log in for availability
1 * 100 mL
Product Details & Documents

Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.

Documents

Specifications

Specifications

  • Concentration:
    1000 mg/l
  • Pk:
    100 mL
  • Description:
    Lactate in water

Specifications

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.