Order Entry
Belgium
ContactUsLinkComponent
Chlorate standard for IC, TraceCERT®, Supelco®
Chlorate standard for IC, TraceCERT®, Supelco®
Catalog # 73166-100ML
Supplier:  Merck
CAS Number:  
Chlorate standard for IC, TraceCERT®, Supelco®
Catalog # 73166-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/l
  • Pk:
    100 mL
  • Description:
    Chlorate in water

Specifications

About this item

Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.