ContactUsLinkComponent
Adipate standard for IC, TraceCERT®, Supelco®
Adipate standard for IC, TraceCERT®, Supelco®
  40968-100ML
 :  Merck
 :  
undefined
Adipate standard for IC, TraceCERT®, Supelco®
  40968-100ML
 :  Merck
 :  
  • Concentration:
    1000 mg/l
  • Pk:
    100 mL
  • Description:
    Adipate in water
 : This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.