Order Entry
Belgium
ContactUsLinkComponent
Tin Standard for ICP, TraceCERT®, Supelco®
Tin Standard for ICP, TraceCERT®, Supelco®
Catalog # 42991-100ML
Supplier:  Merck
CAS Number:  
undefined
Tin Standard for ICP, TraceCERT®, Supelco®
Catalog # 42991-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    5% HNO₃/<0.5% HF
  • Pk:
    100 mL
  • Element:
    Tin
  • Symbol:
    Sn

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034