Order Entry
Belgium
ContactUsLinkComponent
Sulphur standard for AAS, TraceCERT®, Supelco®
Sulphur standard for AAS, TraceCERT®, Supelco®
Catalog # 18020-250ML
Supplier:  Merck
CAS Number:  
Sulphur standard for AAS, TraceCERT®, Supelco®
Catalog # 18020-250ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    250 mL
  • Environmentally Preferable:
  • M-Clarity quality level:
    MQ300
  • Element:
    Sulphur
  • Symbol:
    S

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034