Order Entry
Belgium
ContactUsLinkComponent
 
Silver standard for ICP-MS, TraceCERT®, Supelco®
Catalog # 69389-100ML
Supplier:  Merck
CAS Number:  
Silver standard for ICP-MS, TraceCERT®, Supelco®
Catalog # 69389-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1 mg/L
  • Matrix:
    Nitric acid dil.
  • Pk:
    100 mL
  • Environmentally Preferable:
  • Element:
    Silver
  • Symbol:
    Ag

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034