Order Entry
Belgium
ContactUsLinkComponent
Scandium atomic absorption standard solution, analytical standard, Supelco®
Scandium atomic absorption standard solution, analytical standard, Supelco®
Catalog # 207462-500ML
Supplier:  Merck
CAS Number:  
Scandium atomic absorption standard solution, analytical standard, Supelco®
Catalog # 207462-500ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    1% HNO₃
  • Pk:
    500 mL
  • Element:
    Scandium
  • Symbol:
    Sc

Specifications

About this item

Exact Sc concentration on label.