Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Phosphorus standard for AAS, TracecERT®, Supelco®

Phosphorus, P, 1000 mg/L, H₂O, 250 mL
Catalog # 51474-250ML
Supplier:   Merck
Phosphorus standard for AAS, TracecERT®, Supelco®
Catalog # 51474-250ML
Supplier:  Merck
CAS Number:  
Phosphorus standard for AAS, TracecERT®, Supelco®

Phosphorus, P, 1000 mg/L, H₂O, 250 mL

CAS Number:  
Price
39,40 €
Log in for availability
1 * 250 mL
Product Details & Documents

 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034

 

Specifications

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    250 mL
  • Element:
    Phosphorus
  • Symbol:
    P
  • Cat. No.:
    51474-250ML
  • M-Clarity quality level:
    MQ300

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.