Order Entry
Belgium
Orders LinkContactUsLinkComponent
Multi-element standard solution 5 for ICP, TraceCERT®, Supelco®

Multi-element standard solution 5 for ICP, TraceCERT®, Supelco®

Multi-element standard solution 5 for ICP, 10% HNO₃, 100 mL
  54704-100ML
 :   Merck
Multi-element standard solution 5 for ICP, TraceCERT®, Supelco®
  54704-100ML
 :  Merck
 :  
Multi-element standard solution 5 for ICP, TraceCERT®, Supelco®
Multi-element standard solution 5 for ICP, TraceCERT®, Supelco®

Multi-element standard solution 5 for ICP, 10% HNO₃, 100 mL

 :  
 
181,00 €
   
1 * 100 mL
 

27 components:

Ag, 10 mg/l; Al, 10 mg/l; Ba, 10 mg/l; Be, 10 mg/l; Bi, 10 mg/l
Ca, 100 mg/l; Cd, 10 mg/l; Co, 10 mg/l; Cr, 10 mg/l; Cs, 10 mg/l; Cu, 10 mg/L
Fe, 100 mg/l; Ga, 10 mg/l; In, 10 mg/l; K, 100 mg/l; Li, 10 mg/l
Mg, 10 mg/l; Mn, 10 mg/l; Mo, 10 mg/l; Na, 100 mg/l; Ni, 10 mg/l
Pb, 10 mg/l; Rb, 10 mg/l; Sr, 10 mg/l; Tl, 10 mg/l; V, 10 mg/l; Zn, 10 mg/l

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034

Documents

 

 

  • Matrix:
    10% HNO₃
  • Pk:
    100 mL
  • Description:
    Multi-element standard solution 5 for ICP
  • Cat. No.:
    54704-100ML
  • M-Clarity quality level:
    MQ300

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.