Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Multi-element standard solution 1 for ICP, TraceCERT®, Supelco®

Multi-element standard solution 1 for ICP, 10% HNO₃, 100 mL
  90243-100ML
 :   Merck
Multi-element standard solution 1 for ICP, TraceCERT®, Supelco®
  90243-100ML
 :  Merck
 :  
Multi-element standard solution 1 for ICP, TraceCERT®, Supelco®
Multi-element standard solution 1 for ICP, TraceCERT®, Supelco®

Multi-element standard solution 1 for ICP, 10% HNO₃, 100 mL

 :  
 
166,00 €
   
1 * 100 mL
 

Multi-element standard solution 1 has been used in the analysis of pollutants like trace elements in water using total reflection x-ray fluorescence (TXRF) method and inductively coupled plasma mass spectrometry(ICP-MS).

22 components:

Ag, 10 mg/l; Al, 50 mg/l; B, 50 mg/l; Ba, 10 mg/l; Bi, 100 mg/l
Ca, 10 mg/l; Cd, 10 mg/l; Co, 10 mg/l; Cr, 50 mg/l; Cu, 10 mg/l
Fe, 10 mg/l; K, 100 mg/l; Li, 50 mg/l; Mg, 10 mg/l; Mn, 10 mg/l
Mo, 50 mg/l; Na, 50 mg/l; Ni, 50 mg/l; Pb, 100 mg/l; Sr, 10 mg/l
Tl, 50 mg/l; Zn, 10 mg/l

 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034

 

 

 

  • Matrix:
    10% HNO₃
  • Pk:
    100 mL
  • Description:
    Multi-element standard solution 1 for ICP
  • Cat. No.:
    90243-100ML
  • M-Clarity quality level:
    MQ300

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.