ContactUsLinkComponent
Multi-element Ion chromatography cation standard solution, certified, Supelco®
Multi-element Ion chromatography cation standard solution, certified, Supelco®
Catalog # 89316-50ML-F
Supplier:  Merck
CAS Number:  
Multi-element Ion chromatography cation standard solution, certified, Supelco®
Catalog # 89316-50ML-F
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    50 mL
  • Description:
    Multi-element Ion Chromatography Cation Standard Solution
  • M-Clarity quality level:
    MQ300
Specifications
About this item
10,0 mg/kg ±0,2% Li+, Na+, K+, Mg²⁺, Ca²⁺ each cation, certified reference material traceable to SI.