Order Entry
Belgium
ContactUsLinkComponent
Molybdenum standard for AAS, TraceCERT®, Supelco®
Molybdenum standard for AAS, TraceCERT®, Supelco®
Catalog # 67210-250ML
Supplier:  Merck
CAS Number:  
Molybdenum standard for AAS, TraceCERT®, Supelco®
Catalog # 67210-250ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HCl
  • Pk:
    250 mL
  • M-Clarity quality level:
    MQ300
  • Element:
    Molybdenum
  • Symbol:
    Mo

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034