Cobalt Standard for ICP-MS, TraceCERT®, Supelco®
N° de catalogue 41798-100ML
Fournisseur: Merck
Numéro CAS:
Spécifications
- Concentration:1 mg/L
- Matrix:Nitric acid dil.
- Pk:100 mL
- Element:Cobalt
- Symbol:Co
Spécifications
À propos de cet article
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034