Order Entry
Belgium
ContactUsLinkComponent
Boron standard for ICP, TraceCERT®, Supelco®
Boron standard for ICP, TraceCERT®, Supelco®
Catalog # 18822-100ML
Supplier:  Merck
CAS Number:  
Boron standard for ICP, TraceCERT®, Supelco®
Catalog # 18822-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    <0.5% NH₄OH
  • Pk:
    100 mL
  • Element:
    Boron
  • Symbol:
    B

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034