Order Entry
Belgium
ContactUsLinkComponent
Arsenic standard for ICP-MS, TraceCERT®, Supelco®
Arsenic standard for ICP-MS, TraceCERT®, Supelco®
  75016-100ML
 :  Merck
 :  
Arsenic standard for ICP-MS, TraceCERT®, Supelco®
  75016-100ML
 :  Merck
 :  

 

  • Concentration:
    1 mg/L
  • Matrix:
    Nitric acid dil.
  • Pk:
    100 mL
  • Element:
    Arsenic
  • Symbol:
    As

 

 

 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034