Order Entry
Belgium
ContactUsLinkComponent
 
Corrosive
Environment
Silver sulphate 80 g/l in sulphuric acid 96%, AVS TITRINORM for COD determination according to DIN 38409-H 43-1
Catalog # 86189.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Environment
Silver sulphate 80 g/l in sulphuric acid 96%, AVS TITRINORM for COD determination according to DIN 38409-H 43-1
Catalog # 86189.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Composition complies with:
    DIN 38409 part 43 - DEV H43

Specification Test Results

About this item

 :
 :311.8 g/mol
UN:3264
ADR:8,II
 :GHS05
 :GHS09
CAS Number:10294-26-5
Formula:Ag₂SO₄
MDL Number:MFCD00003407