Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 0.02 mol/l (0.02 N), AVS TITRINORM
Sodium hydroxide 0.02 mol/l (0.02 N), AVS TITRINORM
Catalog # 86188.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium hydroxide 0.02 mol/l (0.02 N), AVS TITRINORM
Catalog # 86188.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :40 g/mol
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548