Order Entry
Belgium
ContactUsLinkComponent
Potassium dihydrogen phosphate 1/15 mol/l (1/15 M), AVS TITRINORM buffer stock solution
Potassium dihydrogen phosphate 1/15 mol/l (1/15 M), AVS TITRINORM buffer stock solution
Catalog # 86185.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium dihydrogen phosphate 1/15 mol/l (1/15 M), AVS TITRINORM buffer stock solution
Catalog # 86185.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Potassium phosphate, monobasic:
    0.0664 – 0.0670 mol/l

Specification Test Results

About this item

 :
 :136.09 g/mol
CAS Number:7778-77-0
Formula:KH₂PO₄
MDL Number:MFCD00011401