Order Entry
Belgium
ContactUsLinkComponent
Disodium hydrogen phosphate solution 1/15 mol/l (1/15 M), AVS TITRINORM buffer stock solution
Disodium hydrogen phosphate solution 1/15 mol/l (1/15 M), AVS TITRINORM buffer stock solution
Catalog # 86184.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Disodium hydrogen phosphate solution 1/15 mol/l (1/15 M), AVS TITRINORM buffer stock solution
Catalog # 86184.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Sodium phosphate, dibasic, anhydrous:
    0.0664 – 0.0670 mol/l

Specification Test Results

About this item

 :
 :141.96 g/mol
CAS Number:7558-79-4
Formula:Na₂HPO₄
MDL Number:MFCD00003496