Order Entry
Belgium
ContactUsLinkComponent
Potassium dichromate solution 0.02 mol/l (0.12 N), AVS TITRINORM for COD determination
Toxic
Health
Potassium dichromate solution 0.02 mol/l (0.12 N), AVS TITRINORM for COD determination
Catalog # 86181.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Health
Potassium dichromate solution 0.02 mol/l (0.12 N), AVS TITRINORM for COD determination
Catalog # 86181.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :294.18 g/mol
Storage Temperature:Ambient
UN:3287
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:7778-50-9
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367