Phosphoryl trichloride ≥99.0% (by argentometric titration), GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10025-87-3
:
Formula:POCl₃
MDL Number:MFCD00011443
:153.33 g/mol
EINECS:233-046-7
UN:1810
ADR:8,I
:GHS06
:GHS05
:GHS08
Specification Test Results
- Appearance:Clear colourless to yellowish liquid
- Assay:≥99.0%
- Colour (APHA):≤30
- Identity:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number Frequently Bought Together